missing translation for 'onlineSavingsMsg'
Learn More

CUDC-427, MedChemExpress

Product Code. 16442927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442927

Brand: Medchem Express HY15835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CUDC-427 is a potent second-generation pan-selective IAP antagonist, used for treatment of various cancers.

CUDC-427 is a potent second-generation pan-selective IAP antagonist, used for treatment of various cancers.

Specifications

Chemical Name or Material CUDC-427
CAS 1446182-94-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H36N6O4S
Quantity 50 mg
Synonym GDC-0917
Solubility Information DMSO : ≥ 100 mg/mL (177.09 mM)
SMILES O=C([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)NC3=C(C4=CC=CC=C4)N=C(C5=NC=CO5)S3
Molecular Weight (g/mol) 564.7
Formula Weight 564.7
Percent Purity 96.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.