missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
Brand: Invitrogen PV3838
Additional Details : Weight : 0.00900kg
Description
The casein kinase I (CKI) gene family encodes serine⁄threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cellsSpecifications
NP_004375, R174G | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
78.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
CSNK1G3 (CK1 γ 3) | |
CSNK1G3 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β−glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200 μM ATP, 330μg/mL Casein Kinase 1 peptide substrate (RRKDLHDDEEDEAMSITA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.36mg/mL | |
1456, 4090 | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
–80°C | |
1 Each | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
CK1 Kinase family | |
160nmoles of phosphate transferred to Casein Kinase 1 peptide substrate (RRKDLHDDEEDEAMSITA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
Kinases and inhibitors | |
80% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.