missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
674.00 EUR - 1826.00 EUR
Specifications
Accession Number | NP_004375, R174G |
---|---|
Concentration | 0.36mg/mL |
Molecular Weight (g/mol) | 78.9kDa |
Name | CSNK1G3 (CK1 γ 3) Recombinant Human Protein |
Storage Requirements | –80°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11503577
|
Invitrogen
PV3838 |
10μg |
674.00 EUR
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15485030
|
Invitrogen
PR7474A |
100μg |
1826.00 EUR
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The casein kinase I (CKI) gene family encodes serine⁄threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cellsSpecifications
NP_004375, R174G | |
78.9kDa | |
–80°C | |
1 Each | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
160nmoles of phosphate transferred to Casein Kinase 1 peptide substrate (RRKDLHDDEEDEAMSITA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
80% by SDS-PAGE |
0.36mg/mL | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
For research use only. Not for use in diagnostic procedures | |
CSNK1G3 (CK1 γ 3) | |
CSNK1G3 (CK1 γ 3) Recombinant Human Protein | |
Human | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.