Learn More
Thermo Scientific™ Cryopreservation Vials, Canes and Canisters
Shop All Thermo Scientific Products
Click to view available options
Type:
Cryo Cane
For Use With (Equipment):
12.5 mm dia. Cryo Tubes
2 Ampules
6 Ampules
9.2 and 10 mL Goblets
Unit Size:
6 ampoules
Each
Description
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
Maximize your cryopreservation storage capacities with a range of vials, canes and canisters.
Specifications
Specifications
| Description | Aluminum |
| Type | Cryo Cane |
| Capacity (Metric) | 5 mL |
| Color | Silver |
| For Use With (Equipment) | 2 Ampules |
| Holds | Ampules (2) |