Learn More
SPEX Certiprep™ CryoPlate aluminum
Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
SPEX GenoGrinder 2010 Homogenizer
Unit Size:
Each
Description
- The 2189C cryoplate keeps samples cold during the grinding process in the 2010 Geno/Grinder™.
- It should be used with the 2189 adjustable clamp.
- To use, simply place the cryoplate on top of vials or titer plates loaded in the adjustable clamp. Fasten the adjustable clamp then start grinding process.
- The cryoplate can also be used in conjunction with cryo-blocks for vials or the 2650 cryo-adapter for titer plates.
Specifications
Specifications
| For Use With (Equipment) | SPEX GenoGrinder 2010 Homogenizer |
| Material | Aluminum |