Learn More
Fisherbrand™ Cryogenic Labels for Cryogenic Storage
GREENER_CHOICE
Product Code. 17847281 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Purple
Red
White
Yellow
Quantity:
1000/Roll
5000/Pk.
For Use With (Equipment):
0.2mL PCR Tubes
0.5 to 1.5mL tube
1.5 and 2 mL tubes
1.5 to 2 mL tubes
15 and 50 mL tube
Size (English):
0.25 x 0.75 in.
0.375 in. dia
0.375 in. dia.
0.5 in. dia
0.5 x 1.25 in.
0.75 in. dia.
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity For Use With (Equipment) Size (English) unitSize
17847281 Purple 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
17807281 Blue 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
17877281 White 5000/Pk. 0.5 to 1.5mL tube 0.375 in. dia. Each
17857281 White 1000/Roll 0.2mL PCR Tubes 0.25 x 0.75 in. Each
17827281 Green 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
17807291 White 1000/Roll 15 and 50 mL tube 0.75 in. dia. Each
17867281 White 1000/Roll 0.5 to 1.5mL tube 0.375 in. dia. Each
17897271 White 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
17897281 White 1000/Roll 1.5 and 2 mL tubes 0.5 in. dia Each
17887281 Assorted 5000/Pk. 0.5 to 1.5mL tube 0.375 in. dia Each
17837281 Yellow 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
17817281 Red 1000/Roll 1.5 to 2 mL tubes 0.5 x 1.25 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 17847281
Only null left
Add to basket
Add to basket
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy-to-use dispenser box with 20 sheets per pack.
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to +121°C (-321° to +250°F)
  • No need to self-laminate
  • Thinner labels allow tubes to fit most storage systems
  • Available in a full range of colors and sizes
  • Guaranteed not to fall off under normal use (provided user instructions are followed)
  • Also offered in laser printer-compatible sheets (see Cat. Nos. 15-930 and 15-940 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifications
Color Purple
Format Label
Product Type Cryogenic Label
For Use With (Application) Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
For Use With (Equipment) 1.5 to 2 mL tubes
Quantity 1000/Roll
Size (English) 0.5 x 1.25 in.
Size (Metric) 1.2 x 3.1 cm
Temperature Range (English) -321 to 250°F
Temperature Range (Metric) -196°C to +121°C
Dimensions (L x W) 0.5 in. x 1.25 in.