missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ White Cryobox with Dividers, 32 mm

Product Code. 11501705 Shop All TENAK Products
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11501705 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11501705 Supplier Tenak™ Supplier No. TE22225
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Cryobox with dividers for cryogenic storage.

  • Cryobox
  • For use with cryogenic storage applications

Specifications

Color White
Includes 9 x 9 Dividers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.