missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIP1 Polyclonal Antibody

CRIP1 Polyclonal Antibody
Brand: Invitrogen PA596715
This item is not returnable.
View return policy
Description
CRIP1 Polyclonal Antibody for Western Blot
Specifications
| CRIP1 | |
| Polyclonal | |
| Unconjugated | |
| CRIP1 | |
| CRHP; Crip; CRIP1; CRP1; CRP-1; cysteine rich intestinal protein; cysteine rich protein 1; cysteine-rich heart protein; Cysteine-rich intestinal protein; cysteine-rich protein 1; cysteine-rich protein 1 (intestinal) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12925, 1396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50238, P63254 | |
| CRIP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-77 of human CRIP1 (NP_0013021). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction