missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPI-637, MedChemExpress
Description
CPI-637 is a selective and potent CBP/EP300 bromodomain inhibitor with IC50 values of 0.03 μM, 0.051 μM and 11.0 μM for CBP, EP300 and BRD4 BD-1, respectively, and an EC50 of 0.3 μM for CBP.
Specifications
Specifications
| Chemical Name or Material | CPI-637 |
| Physical Form | Solid |
| CAS | 1884712-47-3 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22N6O |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 9.62 mg/mL (24.89 mM; Need ultrasonic) |
| SMILES | O=C1NC2=CC=CC(C3=CC4=C(N(C)N=C4C5=CN(C)N=C5)C=C3)=C2N[C@H](C)C1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?