Learn More
Cole-Parmer™ CPC™ All-Plastic Quick Disconnect Barbed Inserts
Click to view available options
Packaging Quantity:
5/Pk.
Unit Size:
Pack of 5
Description
- All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately).
- Simply press the thumb latch to disconnect fitting halves.
- Available in polycarbonate (PC), Acetal and polysulfone (PSF).
Specifications
Specifications
| Type | All-Plastic Quick Disconnect Barbed Inserts; PS, 1/2 in. Tube ID |
| Packaging Quantity | 5/Pk. |
| Description | Quick-Disconnect x Barb |
| Sterilization Method | Autoclave, Ethylene Oxide (ETO) or Gamma |
| Material | Polysulfone |