missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CP110 Recombinant Rabbit Monoclonal Antibody (HL2312)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550487
This item is not returnable.
View return policy
Description
CP110 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Necessary for centrosome duplication at different stages of procentriole formation. Acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation (PubMed:17719545 PubMed:17681131, PubMed:23486064). Also involved in promoting ciliogenesis. May play a role in the assembly of the mother centriole subdistal appendages (SDA) thereby effecting the fusion of recycling endosomes to basal bodies during cilia formation. Required for correct spindle formation and has a role in regulating cytokinesis and genome stability via cooperation with CALM1 and CETN2 (PubMed:16760425). [UniProt].
Specifications
| CP110 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| O43303, Q7TSH4 | |
| CCP110 | |
| Recombinant fragmemt of human CP110. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2312 | |
| Unconjugated | |
| CCP110 | |
| 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; centriolar coiled coil protein 110; centriolar coiled coil protein 110kDa; centriolar coiled-coil protein 110; centriolar coiled-coil protein 110kDa; centriolar coiled-coil protein of 110 kDa; Centriolin; centrosomal protein CP110; centrosomal protein of 110 kDa; CEP1; Cep110; CNTRL; CP110; FAN; Kiaa0419; RGD1310509 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 101565, 361634, 9738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction