Learn More
CP-809101 hydrochloride, MedChemExpress
Product Code. 16462707
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16462707
Brand: Medchem Express HY15543A
MedChemExpress CP-809101 hydrochloride is a potent and selective 5-HT2C receptor agonist with pEC50 of 9.96/7.19/6.81 for human 5-HT2C/5-HT2B/5-HT2A receptors respectively.
CP-809101 hydrochloride is a potent and selective 5-HT2C receptor agonist with pEC50 of 9.96/7.19/6.81 for human 5-HT2C/5-HT2B/5-HT2A receptors respectively.
Specifications
Chemical Name or Material CP-809101 hydrochloride
Physical Form Solid
CAS 1215721-40-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H18Cl2N4O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 20 mg/mL (58.61 mM; Need ultrasonic)
SMILES [H]Cl.ClC1=CC(COC2=NC(N3CCNCC3)=CN=C2)=CC=C1
Molecular Weight (g/mol) 341.24
Formula Weight 341.24
Percent Purity 99.83%
Grade Research