Learn More
Hirschmann™ Cover Glasses for Microscopy
Click to view available options
Dimensions (L x W):
15 x 15 mm
18 x 18 mm
20 x 20 mm
22 x 22 mm
22 x 32 mm
22 x 40 mm
22 x 50 mm
22 x 60 mm
24 x 24 mm
24 x 32 mm
24 x 46 mm
24 x 48 mm
24 x 50 mm
Unit Size:
Pack of 1000
Description
- 0.13-0.17 mm thickness
- Acc. to DIN 58 884, CE acc. IVD 98/79 EC
- Provided in packs of 1000
- With lot number on packaging
Specifications
Specifications
| Product Type | Coverslip |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Dimensions (L x W) | 22 x 22 mm |
| Certifications | CE, DIN 58884, IVD 98/79 EC |