Learn More
Hirschmann™ Counting Chambers
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Nageotte)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Counting Chamber (Thoma-neu)
Description:
Double Net Ruling, Bright Lined
Double Net Ruling, Clamps
Double Net Ruling, Clamps, Bright Lined
Unit Size:
Each
Description
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Specifications
Specifications
| Product Type | Counting Chamber (Nageotte) |
| Description | Double Net Ruling, Bright Lined |
| Quantity | 1 Each |
| Depth (Metric) | 0.5 mm |
| Certifications/Compliance | CE acc. IVD 98/79 EC |
| Includes | 2 cover glasses for haemacytometer |