Learn More
Cotinine N-Beta-D-Glucuronide, TRC
Product Code. 17961485
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961485 1 mg 1mg
17910325 10 mg 10mg
17911285 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17961485 Supplier Toronto Research Chemicals Supplier No. TRCC7251751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 139427-57-9
Molecular Formula C16 H20 N2 O7
Molecular Weight (g/mol) 352.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S)-1-methyl-5-oxo-2-pyrrolidinyl]-, inner salt, Pyridinium, 1-β-D-glucopyranuronosyl-3-(1-methyl-5-oxo-2-pyrrolidinyl)-, inner salt, (S)-, Cotinine-N-glucuronide, Cotinine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[3-[(3S)-1-methyl-5-oxopyrrolidin-3-yl]pyridin-1-ium-1-yl]oxane-2-carboxylate
SMILES CN1C[C@@H](CC1=O)c2ccc[n+](c2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C16H20N2O7/c1-17-6-9(5-10(17)19)8-3-2-4-18(7-8)15-13(22)11(20)12(21)14(25-15)16(23)24/h2-4,7,9,11-15,20-22H,5-6H2,1H3/t9-,11+,12+,13-,14+,15-/m1/s1
Formula Weight 352.1271
Percent Purity >95%
Chemical Name or Material Cotinine N-beta-D-Glucuronide
Grade HPLC