missing translation for 'onlineSavingsMsg'
Learn More

Cotinine N-Beta-D-Glucuronide, TRC

Product Code. 17910325
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910325 10 mg 10mg
17961485 1 mg 1mg
17911285 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17910325 Supplier Toronto Research Chemicals Supplier No. TRCC72517510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139427-57-9
Molecular Formula C16 H20 N2 O7
Molecular Weight (g/mol) 352.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S)-1-methyl-5-oxo-2-pyrrolidinyl]-, inner salt, Pyridinium, 1-β-D-glucopyranuronosyl-3-(1-methyl-5-oxo-2-pyrrolidinyl)-, inner salt, (S)-, Cotinine-N-glucuronide, Cotinine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[3-[(3S)-1-methyl-5-oxopyrrolidin-3-yl]pyridin-1-ium-1-yl]oxane-2-carboxylate
SMILES CN1C[C@@H](CC1=O)c2ccc[n+](c2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H20N2O7/c1-17-6-9(5-10(17)19)8-3-2-4-18(7-8)15-13(22)11(20)12(21)14(25-15)16(23)24/h2-4,7,9,11-15,20-22H,5-6H2,1H3/t9-,11+,12+,13-,14+,15-/m1/s1
Formula Weight 352.1271
Percent Purity >95%
Chemical Name or Material Cotinine N-beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.