Description
- Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
- Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
- Each carrier kit contains eight carriers and eight handles
Specifications
Specifications
| Type | Tray |
| No. per Pack | 25/Pk. |
| No. per Case | 200/Cs. |
| Color | Black |