Learn More
Corning™ Costar™ Netwell™ Inserts
GREENER_CHOICE
Product Code. 10337701 Shop All Corning Products
Change view
Click to view available options
Pore Size:
74 μm
Unit Size:
Pack of 48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10337701 74 μm Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10337701 Supplier Corning™ Supplier No. 3479
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
TRUSTED_SUSTAINABILITY
Specifications
Array 2 x 3
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Cell Culture Insert
Well Count 6
For Use With (Application) Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
No. per Case 48/Cs.
No. per Pack 6/Pk.
Packaging 4 inner packs of 12
Pore Size 74 μm
Quantity 48/Cs.