missing translation for 'onlineSavingsMsg'
Learn More

Anti-coronavirus (SARS-CoV-2) spike S2, mouse, mAb, MP Biomedicals™

Monoclonal antibody raised against the coronavirus spike S2 protein, a common target for neutralizing antibodies and vaccines.

213.00 EUR - 443.00 EUR

Specifications

Antigen Coronavirus Spike S2 Protein
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Host Species Mouse
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16592518
View Documents
MP Biomedicals
08720401
50 μg
ONLINE EXCLUSIVE PRICE
242.00 EUR
213.00 EUR / 50µg
Save 29.00 EUR
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16502528
View Documents
MP Biomedicals
08720402
250 μg
ONLINE EXCLUSIVE PRICE
503.00 EUR
443.00 EUR / 250µg
Save 60.00 EUR
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Anti-coronavirus (SARS-CoV-2) spike S2, mouse, mAb. The 2019 Novel Coronavirus (SARS-CoV-2) is a coronavirus identified as the cause of an outbreak of respiratory illness first detected in Wuhan, China. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M), and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.

Specifications

Specifications

Coronavirus Spike S2 Protein
Monoclonal
Mouse
None
Primary
Western Blot
Unconjugated
RUO
IgG1
Videos
SDS
Documents

Documents

Product Certifications

For research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.