missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Coronavirus Ig Total Panel, 11plex

Product Code. 17156448
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17156448 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17156448 Supplier Invitrogen™ Supplier No. EPX11016000901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ProcartaPlex Human Coronavirus Ig Total Panel 11plex enables screening of four anti-SARS-CoV-2 antibodies, six anti-coronavirus strains, and one negative control in a single well using Luminex xMAP technology.

The ProcartaPlex Human Coronavirus Ig Total Panel 11plex enables screening of four anti-SARS-CoV-2 antibodies (Spike trimer, S1 subunit, RBD (receptor binding domain), and Nucleocapsid), six anti-coronavirus strains (SARS-CoV-1, MERS, CoV-NL63, CoV-KHU1, CoV-229E, CoV-OC43 ), and one negative control in a single well using Luminex xMAP technology. Simultaneous detection of anti-SARS-CoV-2 antibodies and related coronavirus antibodies in one assay can save time to provide a complete, holistic data set of serum samples.

Unlike most ProcartaPlex kits that utilize matched antibody pairs to detect proteins of interest, this serological assay is designed with specific SARS-CoV-2 proteins conjugated to the bead. The detector antibody is an Immunoglobulin (Ig) Total antibody directly conjugated to phycoerythrin (PE) that cross-reacts with all Ig (IgG, IgM, and IgA) in serum. IgG is the most abundant of the immunoglobulins. The panel is provided in a ready-to-use format and the total incubation time is 2.5 hours.

The kit comes with two vials each of high, medium, and low controls to be run either as controls or the high control can be serially diluted to generate a standard curve. Serum and plasma samples are recommended to be diluted 1:1000. It is not combinable with simplexes or other panels.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Target list [bead region]: SARS-CoV-2 Spike (trimer) [14], SARS-CoV-2 S1 protein [15], SARS-CoV-2 RBD (IAP) [75], SARS-CoV-2 Nucleocapsid [13], SARS S1 [67], CoV-NL63 [39], CoV-HKU1 [30], CoV-229E [26], MERS S1 [46], CoV-OC43 [37], Negative control [52]

About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
Well-established Luminex technology—the most referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids. The Luminex MagPlex superparamagnetic microsphere beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create 100 spectrally unique signatures that can be identified by the Luminex xMAP detection systems (Luminex 200, FLEXMAP 3D, and MAGPIX systems).

Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a magnetic bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See product documentation
Cross Reactivity IgG, IgM, IgA
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
Interassay CV Less than 15%
Intraassay CV Less than 15%
Multiplex Size Plex No. 11
Product Line ProcartaPlex
Sample Type Serum, Plasma
Sample Volume Serum, Plasma: 25 μL (prediluted 1:1000)
Content And Storage

• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (50X)
• 2 vials Human Control-High
• 2 vials Human Control-Medium
• 2 vials Human Control-Low
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Assay Buffer (1X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat bottom 96-well plate, black

Store at 2°C to 8°C.

Shipping Condition Wet Ice
Research Area SARS-CoV-2 Ig Total
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.