Learn More
Invitrogen™ CORO7-PAM16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114238
Description
Immunogen sequence: NYEHLFKVND KSVGGSFYLQ SKVVRAKERL DEELKIQAQE DREKGQMPHT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 86%; Human: 100%; Mouse: 86%; Pig: 79%; Rat: 93%.
Specifications
| CORO7-PAM16 | |
| Polyclonal | |
| Unconjugated | |
| CORO7 | |
| 0610011B16Rik; 70 kDa WD repeat tumor rejection antigen homolog; CGI-136; CORO7; coronin 7; coronin-7; Crn7; MAGMAS; magmas-like protein; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; mitochondria-associated granulocyte macrophage CSF-signaling molecule; Mitochondrial import inner membrane translocase subunit TIM16; PAM16; POD1; presequence translocase-associated motor 16 homolog (S. cerevisiae); Presequence translocated-associated motor subunit PAM16; SMDMDM; TIM16; TIMM16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51025, 79585 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P57737, Q9Y3D7 | |
| CORO7, PAM16 | |
| Immunogen sequence: NYEHLFKVND KSVGGSFYLQ SKVVRAKERL DEELKIQAQE DREKGQMPHT. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.