Learn More
Corning™ Vitronectin, Human
Shop All Corning Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Specifications
| For Use With (Application) | Cell Attachment and Proliferation |
| Formulation | Lyophilized (dialyzed against 10 mm Phosphate Buffer pH 7.7); reconstitute in dH2O or Buffered solution at neutral pH |
| Name | Human Vitronectin |
| pH Range | 7.7 |
| Quality Control Testing | >90% by SDS-PAGE |
| Quantity | 0.25 mg |
| Source | Human Plasma |
| Storage Requirements | 2° to 8°C |
| Cross Reactivity | Human |
| Form | Lyophilized |