Learn More
Corning™ Reusable Spinner Flask Accessory, Impeller Assembly
Product Code. 11962011 Shop All Corning Products
Click to view available options
For Use With (Equipment):
Corning Reusable Spinner Flask (36L)
Unit Size:
Each
Product Code. 11962011
Brand: Corning™ 401901
Only null left
Add to basket
Add to basket
For use in magnetically driven, 36L Corning Reusable Spinner Flasks
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 36L Corning™ Reusable Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Corning Reusable Spinner Flask (36L)