Learn More
Corning™ Primaria™ Tissue Culture Flasks
Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
250 mL
Culture Area:
25 cm2
75 cm2
Closure Type:
Plug Seal
Vented
Neck Style:
Canted
Straight
missing translation for 'unitSize'
Pack of 100
Pack of 200
Descripción
- Unique nitrogen-containing tissue culture surface chemistry
- Improves attachment, spreading, and growth for many primary cells or cell lines
- Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
- Stable, permanent surface modification
- Optically clear
- No special storage required
- Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
- Packaged in red color-coded, peel-open, medical-style packages
- Sterilized by gamma irradiation
- Nonpyrogenic tested to less than 0.1 EU/mL
Información de órdenes
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
Especificaciones
Especificaciones
| Closure Color | Blue |
| Closure Material | Phenolic |
| Capacity (Metric) | 50 mL |
| Material | Polystyrene |
| Neck Style | Canted |
| No. per Case | 200/Cs. |
| No. per Pack | 20/Pk. |
| Shape | Rectangular |
| Sterility | Sterile |
| Bottom Shape | Rectangular |
| Mostrar más |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido