Learn More
Corning™ Microvolume Pipet Tips, Sterile
Product Code. 10072521 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 10 μL
Unit Size:
Pack of 960
Product Code. 10072521
Brand: Corning™ 4830
Only null left
Add to basket
Add to basket
Provide accurate, reliable performance in the stated volume range
  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 10 μL
Sterility Sterile
Color Natural
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Microvolume, Round
Packaging Rack
For Use With (Equipment) Eppendorf, Universal Fit Ultramicro