Learn More
Corning™ Microvolume Pipet Tips, Sterile
Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 10 μL
Unit Size:
Pack of 960
Description
- Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Specifications
| Volume (Metric) | 0.1 to 10 μL |
| Sterility | Sterile |
| Color | Natural |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Tip Style | Microvolume, Round |
| Packaging | Rack |
| For Use With (Equipment) | Gilson Pipetman, Universal Fit Ultramicro |
| Material | Polypropylene |