Learn More
Corning™ Clear Polypropylene 384-Well Deep-Well Plates
GREENER_CHOICE
Product Code. 10412952 Shop All Corning Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Product Code. 10412952
Brand: Corning™ 3347
Only null left
Add to basket
Add to basket
Ideal for high-throughput applications requiring added volume
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Packaging Bags of 5
Color Clear
Product Type 384 Deep-Well Microplate
Material Polypropylene
Volume (Metric) Well 240 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
For Use With (Application) For high throughput applications requiring added volume
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.