Learn More
Corning™ Penicillin-Streptomycin Solution
Click to view available options
Concentration:
10,000 IU Penicillin, 10,000μg/mL Streptomycin
5000 IU Penicillin, 5000 μg/mL Streptomycin
Unit Size:
100mL
Pack of 6
Description
- Certificate of Analysis available
- Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis
Specifications
Specifications
| Content And Storage | -20°C |
| Concentration | 5000 IU Penicillin, 5000 μg/mL Streptomycin |
| Form | Solution |
| Product Type | Antibiotic |
| Sterility | Sterile |
| Format | Plastic Bottle |
| Quantity | 6 x 100 mL |