Learn More
Corning™ BioCoat™ Matrigel™ 96-Well, Matrigel-Treated, Flat-Bottom Microplate
Descrizione
Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Specifica
Specifica
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Color | Clear |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Matrigel |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | For culture of polarized cells and angiogenesis assays, promotes capillary tubule formation of endothelial cells and neurite regeneration |
| Quantity | 5/Cs. |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.