Learn More
Corning™ BioCoat™ Control Insert, PET Membrane
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
No. of Wells:
6
24
Inserts per Plate:
6
12
Quantity:
2
4
Unit Size:
Pack of 24
Description
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
Specifications
| Inserts per Plate | 6 |
| Culture Area | 4.2 cm2 |
| Material | PET |
| Pore Size | 1 μm |
| Type | Well Plate |
| Quantity | 4 |
| Storage Requirements | 2° to 8°C |
| No. per Pack | 6/Pk. |
| No. per Case | 24/Cs. |
| Surface Coating | BioCoat Control |
| Show More |