missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PKC Zeta Rabbit Polyclonal Antibody, Proteintech
Shop All Proteintech ProductsDescription
The PKC Zeta antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PKC Zeta. This antibody recognizes human, mouse antigen. The PKC Zeta antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | PKC Zeta |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | PKC-ZETA; PKC2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?