missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated MAP2 Rabbit Polyclonal Antibody, Proteintech
Shop All Proteintech ProductsDescription
The MAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MAP2. This antibody recognizes human, mouse, rat antigen. The MAP2 antibody has been validated for the following applications: IF-P, FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | MAP2 |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | DKFZp686I2148; MAP2A; MAP2B; MAP2C |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?