missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CDC37 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 EUR - 349.00 EUR
Specifications
| Antigen | CDC37 |
|---|---|
| Clone | 1C2A6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237032
|
Proteintech
CL488-66420-20UL |
20 μL |
140.00 EUR 129.00 EUR / 20µL Save 11.00 EUR 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233317
|
Proteintech
CL488-66420-100UL |
100 μL |
450.00 EUR 349.00 EUR / 100µL Save 101.00 EUR 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDC37 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC37. This antibody recognizes human, mouse, rat antigen. The CDC37 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.Specifications
| CDC37 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 11140 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C2A6 | |
| Western Blot | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| P50CDC37 | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit