missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated AMACR/p504S Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60240-100UL
This item is not returnable.
View return policy
Description
The p504S,AMACR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human p504S,AMACR. This antibody recognizes human, mouse antigen. The p504S,AMACR antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| p504S,AMACR | |
| Monoclonal | |
| 1000 μg/mL | |
| CBAS4; RACE | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| 42 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 2H9B5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 23600 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction