missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Copper Reagent Test Kits
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Specifications
| Analytes | 4-Amoniantupyrin |
| For Use With (Application) | For waste water, drinking water, ground water, foodstuffs |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 250 |
| Range | 0.02 - 6.00mg/L |
| Sufficient For | 250 Tests |
| Test Method | Photometric |
| Type | Phenol (C6H5OH) reagent test kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?