Learn More
BINDER™ Copper Interior Fittings
Shop All BINDER Products
Click to view available options
Material:
Copper
Unit Size:
Each
Description
Set Includes:
- 2 shelves (all-copper)
- A water pan (copper-plated)
Specifications
Specifications
| Description | Copper interior fittings - Set consisting of 2 shelves (all-copper) and a water pan (copper-plated) |
| For Use With (Equipment) | BINDER Chambers CB-S170 |
| Material | Copper |
| Type | Fitting |
| Includes | 2 shelves (all-copper) and a water pan (copper-plated) |