Learn More
Corning™ CoolRack™ Plate and Reservoir Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
296.20 EUR - 360.02 EUR
Product Code | Brand | Design | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design | Holds | Price | Quantity & Availability | ||||
15552841
|
Corning™
432070 |
CoolSink XT 96F | One 6-, 12-, 24-, 48-, 96-, or 384-well Flat Bottom Plate, 1 SBS-compatible |
321.52 EUR
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
15562841
|
Corning™
432071 |
CoolSink XT 96U | One 96-well U-bottom Microplate, SBS compatible |
360.02 EUR
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
15572841
|
Corning™
432072 |
CoolSink LX55 | One 55 mL Reagent Reservoir |
296.20 EUR
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
Description
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.Specifications
CoolSink Plate and Reservoir Module |