missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ Plate and Reservoir Modules

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Brand:  Corning™ 432071

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 15562841

  • 360.02 EUR / Each
Estimated Shipment: 04-11-2024
to see stock.
Add to basket

Description

Description

Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • SBS-compatible
  • Specifications

    Specifications

    One 96-well U-bottom Microplate, SBS compatible
    CoolSink XT 96U
    CoolSink Plate and Reservoir Module
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers