Learn More
3M™ Cool Flow™ Fan, 1040 Series, Standard (EU-Plug)
Description
- Reduces heat and moisture build up
- For use with 3M™ 4000+ Series Reusable Half Masks
- Optional attachment that attaches simply and securely
- Noticeable cooling sensation on your face as you work
- Single button control function
- Rechargeable and supplied with EU plug
Specifications
Specifications
| Color | Blue |
| For Use With (Equipment) | 3M™ 4000+ Series Reusable Half Masks |