Learn More
Miele Professional™ Conversion kit UBS 2
Product Code. 15407714 Shop All Miele Products
Click to view available options
:
conversion kit; upper baskets; model G 78 to PG 85
Product Code. 15407714
Brand: Miele Professional™ undefined
Only null left
Add to basket
Add to basket
Conversion kit UBS 2 for use with selected series G 78 upper baskets in PG 85 machines
  • Converts old injector baskets to fit into new Miele PG85 range lab washing machines
  • For load carriers and upper baskets for G 78 model series without drying connection
TRUSTED_SUSTAINABILITY
Specifications
Color Black