Learn More
BINDER™ Convenient Water Supply
Shop All BINDER Products
Click to view available options
Material:
Plastic
Unit Size:
Each
Description
- External
- With magnetic holder
- Flexible positioning of the water canister
- Hose connection to the canister with practical quick coupling and water stop function
Specifications
Specifications
| Description | Convenient water supply - External, with magnetic holder, Flexible positioning of the water container with magnetic holder, hose adapter for water container with practical quick coupling, and water stop function |
| For Use With (Equipment) | BINDER Chambers Series KBF-S |
| Material | Plastic |
| Type | Water Supply |
| Includes | Magnetic Holder |