Learn More
HUBER Control Cables
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
CP AIf / Open
Cc / Rs232 15 Pol
Command Module
Control Cable ECS Standby /Open End
Cp Rs485 / Open
Data Transfer
LEVEL - Open End
PC to Serial Interface
POKO-Alarm/Comgate-Unistat
Unipump/ Unistat
Type:
Control Cable
Control Cable ECS
Control Cable POKO
LEVEL - open end
missing translation for 'unitSize'
Each
Description
Cable Length: 3m
Specifications
Specifications
| For Use With (Equipment) | Control Cable ECS Standby /Open End |
| Type | Control Cable ECS |
| Length (Metric) | 3 m |