Learn More
HUBER Control Cables
Cable Length: 3 m
Brand:  HUBER 55018
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15142698
  • 169.60 EUR / Each
Estimated Shipment: 18-06-2024
Description
Cable Length: 3m
Specifications
Specifications
PC to Serial Interface
Control Cable
Pc To Serial Interface
3 m