Learn More
HUBER Control Cables
Cable Length: 3 m
Brand:  HUBER 6254
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 10262215
  • 95.03 EUR / Each
Estimated Shipment: 18-06-2024
Description
Cable Length: 3m
Specifications
Specifications
CP AIf / Open
Control Cable
3 m