Learn More
SymLine Continuous Level Control Accessories
Click to view available options
Includes:
1 x banana plug, 1 x clamp
1 x clamp, 1 ring ID 10 mm
1 x clamp, 1 x button plug
1 x clamp, 1 x ring ID 5 mm
2 x clamps
2x 10 mm press button
ATEX zone 0, ATEX Ex II 1 G Ex ia IIC T1-T6 Ga
ATEX, II 2 G EEx ia IIC T1-T6, NAMUR
Fitting for continuous level control
Ground cable
Grounding cable and clamp
Product Type:
Antistatic Floor Mat
Capacitive Disc Sensor
Capacitive Rod Sensor
Earthing Cable
Grounding Cable
Grounding Plug
Grounding Strap
Safety Waste Cap
Unit Size:
Each
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications
Specifications
| Description | Grounding strap, rigid, with grounding cable and clamp, suitable for canister 108420 |
| Length (Metric) | 195 mm |
| Quantity | 1 Ea. |
| Includes | Grounding cable and clamp |
| For Use With (Equipment) | Canisters with S90 Thread |
| Material | Stainless Steel |
| Product Type | Grounding Strap |