Learn More
Univet™ Contemporary Clear Safety Glasses
Click to view available options
Color:
Amethyst
Honey
Stone
Size:
Medium
Large
Unit Size:
Each
Pack of 10
Description
- Light and very resistant
- Polyamide frame with zero metallic components
- Integrated side-shields
Specifications
Specifications
| Shape | Rectangular |
| Lens Coating | Anti-fog, Scratch-resistant |
| Color | Stone |
| Lens Tint | None |
| Product Type | Safety Glasses |
| Size | Large |
| Material | Plastic |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polyamide |
| Show More |