missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 5 Monoclonal Antibody (4C10E3C12), Invitrogen™

Description
Specificity: Human CNTN5/Contactin-5
The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
| Antigen | Contactin 5 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4C10E3C12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CNTN5 |
| Gene Accession No. | O94779 |
| Gene Alias | 6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?