missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 5 Monoclonal Antibody (4C10E3C12), Invitrogen™

Product Code. 17769853
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17769853 50 μL 50µL
17779543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17769853 Supplier Invitrogen™ Supplier No. MA543508

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human CNTN5/Contactin-5

The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 5
Applications ELISA
Classification Monoclonal
Clone 4C10E3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CNTN5
Gene Accession No. O94779
Gene Alias 6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2
Gene Symbols CNTN5
Host Species Mouse
Immunogen Recombinant Human CNTN5 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53942
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.