Learn More
Tork Constant Air Freshener Refill A3
Shop All Tork Products
Click to view available options
Scent:
Breeze
Floral
Lemongrass
Variety Pack
Unit Size:
Pack of 6
Description
- For a fresh washroom at all times, this scent contains an odour neutraliser to eliminate smells, not just mask them.
- This propellant-free system uses only pure perfume oil, isn't flammable, and is not pressurised.
- Use with Tork Constant Air Freshener Dispensers for long-lasting, constant scent dispersal.
Refill packs available:
- Blossom scent with delicate floral notes for an inviting feel.
- Breeze scent with ylang-ylang, floral and woody notes for a fresh feel.
- Odour neutraliser scent with a very subtle, unobtrusive lemongrass smell.
- Variety pack includes 3 each of Blossom and Breeze.
Specifications
Specifications
| Product Type | Air Freshener Refill |
| Alternate Part Number | 7322541889337 |
| Color-coded | No |
| Form | Spray |
| Scent | Floral |
| Quantity | 6/Pk. |
| Volume (Metric) | 32 mL |
Safety and Handling
Signal Word
- Warning
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- P264-Wash thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.