Learn More
Conductivity Solution, 50000μS.cm-1, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Specifications
Specifications
| Packaging | Bottle |
| Conductivity | 50,000μS/cm |
| Type | Conductivity Solution |
| Quantity | 1000 mL |